Gene Transcripts

Gene Symbol
KIR2DL5A
Entrez Gene ID
57292
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 5A
Gene Aliases
CD158F, KIR2DL5, KIR2DL5.1, KIR2DL5.3
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.740906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020535.3 NP_065396.1 3 231
GenBank mRNA AF204903.1 - 3 231
AF217487.1 - 4 200

Gene Symbol
KIR2DL5B
Entrez Gene ID
553128
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 5B
Gene Aliases
KIR2DL5, KIR2DL5.2, KIR2DLX
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.740906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018081.1 NP_001018091.1 3 176
GenBank mRNA AF271607.1 - 3 176
AF271608.1 - 3 176
AJ544587.1 - 3 177

View Full Product Details