Gene Transcripts

Gene Symbol
RAP1GDS1
Entrez Gene ID
5910
Gene Name
Rap1 GTPase-GDP dissociation stimulator 1
Gene Aliases
GDS1, SmgGDS
Chromosome Location
Chr. 4: 98261376 - 98443861 on Build GRCh38
UniGene ID
Hs.132858
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100426.1 NP_001093896.1 14 1801
NM_001100427.1 NP_001093897.1 14 1798
NM_001100428.1 NP_001093898.1 13 1654
NM_001100429.1 NP_001093899.1 13 1651
NM_001100430.1 NP_001093900.1 12 1525
NM_021159.4 NP_066982.3 14 1798
GenBank mRNA AF215923.1 - 13 1608
AF237413.1 - 13 1608
AK055032.1 - 14 1721
AK295156.1 - 13 1613
AK297543.1 - 11 1348
AK311243.1 - 11 1336
AY376900.1 - 11 1335
BC001816.2 - 14 1705 3010166
BC001851.2 - 14 1705 3010166
BC098269.1 - 14 1659 40001059
BC098334.1 - 13 1509 40001057
BC099708.1 - 13 1509 40001058
BC099845.1 - 13 1512 40001061
BT006837.1 - 13 1608
X63465.1 - 12 1461

View Full Product Details