Gene Transcripts

Gene Symbol
HIST2H3C
Entrez Gene ID
126961
Gene Name
histone cluster 2, H3c
Gene Aliases
H3, H3.2, H3/M, H3F2, H3FM, H3FN
Chromosome Location
Chr. 1: 149840687 - 149841193 on Build GRCh38
UniGene ID
Hs.647745
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021059.2 NP_066403.2 1 270
GenBank mRNA BC015544.1 - 1 270
BC074969.2 - 1 262 30915509
BC130635.1 - 1 264 40146666
BC130637.1 - 1 264 40146668

Gene Symbol
HIST2H3A
Entrez Gene ID
333932
Gene Name
histone cluster 2, H3a
Gene Aliases
H3/n, H3/o
Chromosome Location
Chr. 1: 149852619 - 149853125 on Build GRCh38
UniGene ID
Hs.647745
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005464.2 NP_001005464.1 1 270

Gene Symbol
HIST2H3D
Entrez Gene ID
653604
Gene Name
histone cluster 2, H3d
Gene Aliases
Chromosome Location
Chr. 1: 149813225 - 149813681 on Build GRCh38
UniGene ID
Hs.712062
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123375.2 NP_001116847.1 1 234

View Full Product Details