Gene Transcripts
- Gene Symbol
- C4orf47
- Entrez Gene ID
- 441054
- Gene Name
- chromosome 4 open reading frame 47
- Gene Aliases
- Chromosome Location
- Chr. 4: 185405227 - 185449828 on Build GRCh38
- UniGene ID
- Hs.549398
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114357.1 |
NP_001107829.1
|
1, 2 |
120 |
|
XM_005263019.3 |
XP_005263076.1
|
3, 4 |
473 |
|
XM_011531989.2 |
XP_011530291.1
|
2, 3 |
399 |
|
XM_017008236.1 |
XP_016863725.1
|
3, 4 |
1866 |
|
XM_017008237.1 |
XP_016863726.1
|
4, 5 |
641 |
|
XM_017008238.1 |
XP_016863727.1
|
2, 3 |
1650 |
|
XM_017008239.1 |
XP_016863728.1
|
2, 3 |
1653 |
|
XM_017008240.1 |
XP_016863729.1
|
2, 3 |
402 |
|
XM_017008242.1 |
XP_016863731.1
|
3, 4 |
1932 |
|
GenBank mRNA |
BC150574.1 |
- |
1, 2 |
120 |
|
View Full Product Details