Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000990.4 | NP_000981.1 | 3 | 499 | |
GenBank mRNA | AK311767.1 | - | 3 | 155 | |
BC005326.1 | - | 1 | 155 | 4052417 | |
BC020169.1 | - | 2 | 131 | ||
BC066326.1 | - | 1 | 155 | 5311530 | |
BG687024.1 | - | 189 | |||
BQ058236.1 | - | 3 | 177 | ||
U14968.1 | - | 3 | 149 |