Gene Transcripts

Gene Symbol
PRSS48
Entrez Gene ID
345062
Gene Name
protease, serine 48
Gene Aliases
ESSPL
Chromosome Location
Chr. 4: 151276482 - 151291856 on Build GRCh38
UniGene ID
Hs.651266
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183375.2 NP_899231.2 3 254
XM_011531921.2 XP_011530223.1 2 700
GenBank mRNA BC121109.1 - 3 258

View Full Product Details