Gene Transcripts

Gene Symbol
FCGR2C
Entrez Gene ID
9103
Gene Name
Fc fragment of IgG receptor IIc (gene/pseudogene)
Gene Aliases
CD32, CD32C, CDW32, FCG2, FCRIIC, IGFR2
Chromosome Location
Chr. 1: 161581339 - 161601220 on Build GRCh38
UniGene ID
Hs.656750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201563.5 NP_963857.3 7 1172
NR_047648.1 7 1172
GenBank mRNA X17652.1 - 7 1103

View Full Product Details