Gene Transcripts

Gene Symbol
KRTAP10-11
Entrez Gene ID
386678
Gene Name
keratin associated protein 10-11
Gene Aliases
KAP10.11, KAP18.11, KRTAP18-11, KRTAP18.11
Chromosome Location
Chr. 21: 44646414 - 44647650 on Build GRCh38
UniGene ID
Hs.58076
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198692.2 NP_941965.2 1 1209
GenBank mRNA AF086314.1 - 1 389
BX103563.1 - 1 387

View Full Product Details