Gene Transcripts

Gene Symbol
AUTS2
Entrez Gene ID
26053
Gene Name
autism susceptibility candidate 2
Gene Aliases
FBRSL2, MRD26
Chromosome Location
Chr. 7: 69598475 - 70793068 on Build GRCh38
UniGene ID
Hs.21631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127231.2 NP_001120703.1 2 1189
NM_001127232.2 NP_001120704.1 2 1189
NM_015570.3 NP_056385.1 2 1189
XM_011516010.2 XP_011514312.1 2 1633
XM_011516011.2 XP_011514313.1 2 1633
XM_011516012.2 XP_011514314.1 2 1633
XM_011516013.2 XP_011514315.1 2 1633
XM_011516014.2 XP_011514316.1 2 1633
XM_011516018.2 XP_011514320.1 2 168
XM_017011951.1 XP_016867440.1 2 1633
GenBank mRNA AB007902.2 - 2 750
AF326917.1 - 2 775
BC011643.2 - 2 497
BC064693.1 - 2 1197 30345831
JQ670866.1 - 2 454

View Full Product Details