Gene Transcripts

Gene Symbol
MAPK8IP3
Entrez Gene ID
23162
Gene Name
mitogen-activated protein kinase 8 interacting protein 3
Gene Aliases
JIP-3, JIP3, JSAP1, SYD2, syd
Chromosome Location
Chr. 16: 1706183 - 1770317 on Build GRCh38
UniGene ID
Hs.207763
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040439.1 NP_001035529.1 12 1624
NM_001318852.1 NP_001305781.1 13 1682
NM_015133.4 NP_055948.2 13 1679
XM_006720869.2 XP_006720932.1 13 3112
XM_011522429.2 XP_011520731.1 14 1706
XM_011522430.2 XP_011520732.1 13 1688
XM_011522433.2 XP_011520735.1 9 2329
XM_017023079.1 XP_016878568.1 12 1664
XM_017023081.1 XP_016878570.1 12 1154
GenBank mRNA AB028989.2 - 13 1642
BC137123.1 - 13 1594 9021120
BC137124.1 - 13 1594 9021121
BC144486.1 - 13 1597 9053013
BC150266.1 - 13 1642 9007222

View Full Product Details