Gene Transcripts

Gene Symbol
Rab7
Entrez Gene ID
19349
Gene Name
RAB7, member RAS oncogene family
Gene Aliases
Rab7a
Chromosome Location
Chr. 6: 87999106 - 88045270 on Build GRCm38
UniGene ID
Mm.333233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293652.1 NP_001280581.1 7, 8 966
NM_001293653.1 NP_001280582.1 5, 6 627
NM_001293654.1 NP_001280583.1 6, 7 793
NM_001293655.1 NP_001280584.1 5, 6 706
NM_009005.3 NP_033031.2 5, 6 744
GenBank mRNA AB232599.1 - 4, 5 538
AK005004.1 - 5, 6 706
AK057671.1 - 5, 6 673
AK135659.1 - 6, 7 795
AK139914.1 - 5, 6 697
AK148117.1 - 5, 6 737
AK151078.1 - 5, 6 718
AK152666.1 - 7, 8 941
AK154234.1 - 5, 6 725
AK158971.1 - 5, 6 628
AK159507.1 - 5, 6 673
AK159928.1 - 5, 6 645
AK183086.1 - 2, 3 202
BC004597.1 - 5, 6 662 3584891
BC086793.1 - 5, 6 660 6414012
CT010352.1 - 4, 5 538
X89650.1 - 5, 6 694

View Full Product Details