Gene Transcripts

Gene Symbol
Gm12942
Entrez Gene ID
100039968
Gene Name
predicted gene 12942
Gene Aliases
OTTMUSG00000009332, Zmym6nb
Chromosome Location
Chr. 4: 127126043 - 127129660 on Build GRCm38
UniGene ID
Mm.392655
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099319.2 NP_001092789.1 2 220
GenBank mRNA AK034325.1 - 1 1849
AK156456.1 - 2 202
AK160613.1 - 1 1843

View Full Product Details