Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081364.3 | NP_001074833.3 | 14, 15 | 3628 | |
NM_001128084.2 | NP_001121556.2 | 15, 16 | 3658 | ||
GenBank mRNA | AK122508.1 | - | 1269 | ||
BC076629.1 | - | 14, 15 | 3628 | 30639338 | |
BC150741.1 | - | 15, 16 | 3587 | 9087652 |