Gene Transcripts
- Gene Symbol
- Uba52
- Entrez Gene ID
- 22186
- Gene Name
- ubiquitin A-52 residue ribosomal protein fusion product 1
- Gene Aliases
- D8Ertd21e, Gm1863, Rps27a, Ubb, Ubc
- Chromosome Location
- Chr. 8: 70508266 - 70510367 on Build GRCm38
- UniGene ID
- Mm.297372
- Species
- Mus musculus
- Gene Symbol
- Gm11517
- Entrez Gene ID
- 629750
- Gene Name
- ubiquitin A-52 residue ribosomal protein fusion product 1 pseudogene
- Gene Aliases
- OTTMUSG00000001634
- Chromosome Location
- Chr. 11: 96795332 - 96798156 on Build GRCm38
- UniGene ID
- Mm.390396
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BY726472.1 |
- |
2 |
459 |
|
View Full Product Details