Gene Transcripts

Gene Symbol
Brwd1
Entrez Gene ID
93871
Gene Name
bromodomain and WD repeat domain containing 1
Gene Aliases
5330419I02Rik, D530019K20Rik, G1-403-16, Wdr9, repro5
Chromosome Location
Chr. 16: 95992089 - 96082428 on Build GRCm38
UniGene ID
Mm.240871
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103179.1 NP_001096649.1 4 311
NM_145125.3 NP_660107.2 4 311
NM_176928.1 NP_795902.1 4 311
GenBank mRNA AJ292467.1 - 4 316
AJ292468.1 - 4 316
AK030493.1 - 4 312
AK051870.1 - 4 333
BC056618.1 - 4 409
BC095940.1 - 4 236
BC145265.1 - 4 237

View Full Product Details