Gene Transcripts

Gene Symbol
Slc7a5
Entrez Gene ID
20539
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 5
Gene Aliases
4F2LC, D0H16S474E, LAT1, TA1
Chromosome Location
Chr. 8: 121881146 - 121907686 on Build GRCm38
UniGene ID
Mm.22374
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011404.3 NP_035534.2 5 993
GenBank mRNA AB017189.1 - 5 939
AB023409.1 - 5 921
AK142288.1 - 5 994
AK148134.1 - 5 663
AK192999.1 - 3 153
AK202552.1 - 3 152
AK207202.1 - 3 183
AK215643.1 - 3 182
BC026131.1 - 5 1002 4188854

View Full Product Details