Gene Transcripts

Gene Symbol
Vmn2r65
Entrez Gene ID
100009609
Gene Name
vomeronasal 2, receptor 65
Gene Aliases
ENSMUSG00000070600, V2ro
Chromosome Location
Chr. 7: 84940169 - 84964009 on Build GRCm38
UniGene ID
Mm.389926
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105180.1 NP_001098650.1 4 1298
GenBank mRNA AB194095.1 - 2 351

View Full Product Details