Gene Transcripts
- Gene Symbol
- Vmn2r31
- Entrez Gene ID
- 100042591
- Gene Name
- vomeronasal 2, receptor 31
- Gene Aliases
- Chromosome Location
- Chr. 7: 7383985 - 7399627 on Build GRCm38
- UniGene ID
- Mm.469891
- Species
- Mus musculus
- Gene Symbol
- Vmn2r33
- Entrez Gene ID
- 624512
- Gene Name
- vomeronasal 2, receptor 33
- Gene Aliases
- Chromosome Location
- Chr. 7: 7550967 - 7566786 on Build GRCm38
- UniGene ID
- Mm.469895
- Species
- Mus musculus
- Gene Symbol
- Vmn2r38
- Entrez Gene ID
- 434110
- Gene Name
- vomeronasal 2, receptor 38
- Gene Aliases
- Chromosome Location
- Chr. 7: 9074796 - 9097765 on Build GRCm38
- UniGene ID
- Mm.469895
- Species
- Mus musculus
- Gene Symbol
- Vmn2r44
- Entrez Gene ID
- 434113
- Gene Name
- vomeronasal 2, receptor 44
- Gene Aliases
- EG434113
- Chromosome Location
- Chr. 7: 8367460 - 8383238 on Build GRCm38
- UniGene ID
- Mm.469891
- Species
- Mus musculus
View Full Product Details