Gene Transcripts

Gene Symbol
NBEAL2
Entrez Gene ID
23218
Gene Name
neurobeachin like 2
Gene Aliases
BDPLT4, GPS
Chromosome Location
Chr. 3: 46979683 - 47009704 on Build GRCh38
UniGene ID
Hs.437043
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015175.2 NP_055990.1 7 774
XM_006713072.3 XP_006713135.1 7 771
XM_011533533.2 XP_011531835.1 7 771
XM_017006010.1 XP_016861499.1 7 771
XM_017006011.1 XP_016861500.1 7 627
XM_017006012.1 XP_016861501.1 7 771
XM_017006013.1 XP_016861502.1 7 771
XM_017006014.1 XP_016861503.1 7 627
XM_017006016.1 XP_016861505.1 2 1348
GenBank mRNA AK092871.1 - 7 628
AK131104.1 - 7 614

View Full Product Details