Gene Transcripts

Gene Symbol
Enho
Entrez Gene ID
69638
Gene Name
energy homeostasis associated
Gene Aliases
1110065P19Rik, 2310040A07Rik
Chromosome Location
Chr. 4: 41638144 - 41640302 on Build GRCm38
UniGene ID
Mm.372459
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027147.1 NP_081423.1 2 433
GenBank mRNA AK009710.1 - 2 433
AK050496.1 - 2 232
BC021944.1 - 2 91
BQ554924.1 - 2 458

View Full Product Details