Gene Transcripts

Gene Symbol
Scgb1c1
Entrez Gene ID
338417
Gene Name
secretoglobin, family 1C, member 1
Gene Aliases
Ryd5
Chromosome Location
Chr. 7: 140845565 - 140846769 on Build GRCm38
UniGene ID
Mm.270960
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099742.1 NP_001093212.1 2 235
GenBank mRNA AA462242.1 - 2 208
CB173308.1 - 2 212

View Full Product Details