Gene Transcripts

Gene Symbol
Gucy2d
Entrez Gene ID
14918
Gene Name
guanylate cyclase 2d
Gene Aliases
Chromosome Location
Chr. 7: 98440416 - 98477572 on Build GRCm38
UniGene ID
Mm.389928
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130693.3 NP_001124165.1 4 1330

View Full Product Details