Gene Transcripts
- Gene Symbol
- KAZN
- Entrez Gene ID
- 23254
- Gene Name
- kazrin, periplakin interacting protein
- Gene Aliases
- KAZ
- Chromosome Location
- Chr. 1: 13893387 - 15118048 on Build GRCh38
- UniGene ID
- Hs.368823
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001017999.2 |
NP_001017999.1
|
7, 8 |
1134 |
|
NM_001018000.3 |
NP_001018000.1
|
7, 8 |
1323 |
|
NM_001018001.2 |
NP_001018001.1
|
7, 8 |
1124 |
|
NM_015209.2 |
NP_056024.1
|
7, 8 |
1386 |
|
NM_201628.2 |
NP_963922.2
|
7, 8 |
1386 |
|
XM_005245795.4 |
XP_005245852.1
|
9, 10 |
1641 |
|
XM_011541074.2 |
XP_011539376.1
|
9, 10 |
1648 |
|
XM_011541075.2 |
XP_011539377.1
|
7, 8 |
1304 |
|
XM_011541076.2 |
XP_011539378.1
|
7, 8 |
1449 |
|
XM_011541077.2 |
XP_011539379.1
|
7, 8 |
2428 |
|
XM_011541080.2 |
XP_011539382.1
|
9, 10 |
1641 |
|
XM_017000770.1 |
XP_016856259.1
|
9, 10 |
1636 |
|
XM_017000771.1 |
XP_016856260.1
|
8, 9 |
1309 |
|
XM_017000772.1 |
XP_016856261.1
|
7, 8 |
988 |
|
GenBank mRNA |
AB015329.1 |
- |
6, 7 |
776 |
|
AK293806.1 |
- |
2, 3 |
192 |
|
AY505119.1 |
- |
8, 9 |
1314 |
|
AY505120.1 |
- |
7, 8 |
1323 |
|
AY505121.1 |
- |
7, 8 |
1124 |
|
AY505122.1 |
- |
7, 8 |
1134 |
|
BC035501.2 |
- |
7, 8 |
1322 |
4556194 |
DR003677.1 |
- |
2, 3 |
109 |
|
View Full Product Details