Gene Transcripts
- Gene Symbol
- Rpl29
- Entrez Gene ID
- 19944
- Gene Name
- ribosomal protein L29
- Gene Aliases
- Rpl43
- Chromosome Location
- Chr. 9: 106429432 - 106431568 on Build GRCm38
- UniGene ID
- Mm.288696
- Species
- Mus musculus
- Gene Symbol
- 4931440P22Rik
- Entrez Gene ID
- 71004
- Gene Name
- RIKEN cDNA 4931440P22 gene
- Gene Aliases
- Chromosome Location
- Chr. 3: 65527483 - 65529414 on Build GRCm38
- UniGene ID
- Mm.475465
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK161073.1 |
- |
|
707 |
|
View Full Product Details