Gene Transcripts

Gene Symbol
EHBP1
Entrez Gene ID
23301
Gene Name
EH domain binding protein 1
Gene Aliases
HPC12, NACSIN
Chromosome Location
Chr. 2: 62673851 - 63046487 on Build GRCh38
UniGene ID
Hs.271667
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142614.1 NP_001136086.1 12, 13 2617
NM_001142615.2 NP_001136087.1 13, 14 2850
NM_001142616.1 NP_001136088.1 13, 14 2804
NM_015252.3 NP_056067.2 14, 15 2909
XM_005264225.2 XP_005264282.1 14, 15 2807
XM_005264226.2 XP_005264283.1 14, 15 3164
XM_005264227.1 XP_005264284.1 13, 14 2763
XM_006711971.2 XP_006712034.1 14, 15 2868
XM_011532713.2 XP_011531015.1 14, 15 2950
XM_011532714.2 XP_011531016.1 14, 15 2799
XM_011532715.2 XP_011531017.1 14, 15 2775
XM_011532716.2 XP_011531018.1 14, 15 2868
XM_011532717.1 XP_011531019.1 14, 15 2868
XM_011532718.2 XP_011531020.1 13, 14 2529
XM_017003643.1 XP_016859132.1 14, 15 2765
XM_017003644.1 XP_016859133.1 14, 15 2742
XM_017003645.1 XP_016859134.1 14, 15 2807
XM_017003646.1 XP_016859135.1 14, 15 2870
XM_017003647.1 XP_016859136.1 13, 14 2702
XM_017003648.1 XP_016859137.1 13, 14 2698
XM_017003649.1 XP_016859138.1 13, 14 2777
XM_017003650.1 XP_016859139.1 14, 15 2807
XM_017003651.1 XP_016859140.1 13, 14 2702
XM_017003652.1 XP_016859141.1 13, 14 2763
XM_017003653.1 XP_016859142.1 13, 14 2691
XM_017003654.1 XP_016859143.1 14, 15 2868
GenBank mRNA AB020710.1 - 6, 7 1622
AK000986.1 - 1, 2 134
AK292545.1 - 13, 14 2850
AL833968.1 - 14, 15 2775
AY331186.1 - 14, 15 2804
AY531390.1 - 12, 13 2322
BC067215.1 - 13, 14 2658 6502257

View Full Product Details