Gene Transcripts

Gene Symbol
FKBP15
Entrez Gene ID
23307
Gene Name
FK506 binding protein 15
Gene Aliases
FKBP133, KIAA0674, PPP1R76
Chromosome Location
Chr. 9: 113165036 - 113221361 on Build GRCh38
UniGene ID
Hs.522351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015258.1 NP_056073.1 17 1773
XM_006717018.2 XP_006717081.1 17 1690
XM_006717019.1 XP_006717082.1 16 1621
XM_017014552.1 XP_016870041.1 16 1621
GenBank mRNA AB014574.1 - 17 1700
AK301208.1 - 4 527
AK303074.1 - 16 1621
AK307498.1 - 16 1602
AK307507.1 - 21 2207
BC077732.1 - 1 16

View Full Product Details