Gene Transcripts

Gene Symbol
Cys1
Entrez Gene ID
690489
Gene Name
cystin 1
Gene Aliases
Chromosome Location
Chr. 6: 43844937 - 43862131 on Build Rnor_6.0
UniGene ID
Rn.233945
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109597.3 NP_001103067.1 3 2642

View Full Product Details