Gene Transcripts

Gene Symbol
Prr15
Entrez Gene ID
312358
Gene Name
proline rich 15
Gene Aliases
RGD1311589
Chromosome Location
Chr. 4: 84478839 - 84481885 on Build Rnor_6.0
UniGene ID
Rn.37820
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104527.1 NP_001097997.1 2 640
GenBank mRNA BC127529.1 - 2 640

View Full Product Details