Gene Transcripts
- Gene Symbol
- Mrps21
- Entrez Gene ID
- 689432
- Gene Name
- mitochondrial ribosomal protein S21
- Gene Aliases
- Chromosome Location
- Chr. 2: 197974758 - 197982385 on Build Rnor_6.0
- UniGene ID
- Rn.133208
- Species
- Rattus norvegicus
- Gene Symbol
- Mrps21l
- Entrez Gene ID
- 364906
- Gene Name
- mitochondrial ribosomal protein S21-like
- Gene Aliases
- Mrps21
- Chromosome Location
- Chr. 18: 76317078 - 76317542 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_344706.5 |
XP_344707.1
|
1 |
156 |
|
View Full Product Details