Gene Transcripts

Gene Symbol
Derl3
Entrez Gene ID
690315
Gene Name
derlin 3
Gene Aliases
Chromosome Location
Chr. 20: 13692123 - 13707625 on Build Rnor_6.0
UniGene ID
Rn.20722
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109577.1 NP_001103047.2 5 445
XM_006256325.2 XP_006256387.1 5 1834
XM_006256326.2 XP_006256388.1 5 1831
XM_008772914.1 XP_008771136.1 3 640
XM_008772915.1 XP_008771137.1 3 640
XM_008772916.1 XP_008771138.1 3 640
XM_008772917.1 XP_008771139.1 3 640
XM_008772918.1 XP_008771140.1 5 1837
XM_008772919.1 XP_008771141.1 3 640
XM_008772920.1 XP_008771142.1 5 1834
XM_008772921.1 XP_008771143.1 4 529
XM_008772922.1 XP_008771144.1 4 1772
XM_008772923.1 XP_008771145.1 3 640
XM_008772924.1 XP_008771146.1 3 640
GenBank mRNA BC167034.1 - 5 402

Gene Symbol
LOC103694875
Entrez Gene ID
103694875
Gene Name
derlin-3
Gene Aliases
Chromosome Location
Chr. Un|NW_007906819: 45160 - 60662 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773829.1 XP_008772051.1 3 640
XM_008773830.1 XP_008772052.1 3 640
XM_008773831.1 XP_008772053.1 3 640
XM_008773832.1 XP_008772054.1 3 640
XM_008773833.1 XP_008772055.1 5 1837
XM_008773834.1 XP_008772056.1 3 640
XM_008773835.1 XP_008772057.1 5 1834
XM_008773836.1 XP_008772058.1 4 529
XM_008773837.1 XP_008772059.1 4 1772
XM_008773838.1 XP_008772060.1 3 640
XM_008773839.1 XP_008772061.1 3 640
XM_008773840.1 XP_008772062.1 5 1833
XM_008773841.1 XP_008772063.1 5 1834
XM_008773842.1 XP_008772064.1 5 1831

View Full Product Details