Gene Transcripts

Gene Symbol
Gys1
Entrez Gene ID
690987
Gene Name
glycogen synthase 1, muscle
Gene Aliases
Chromosome Location
Chr. 1: 101427195 - 101447092 on Build Rnor_6.0
UniGene ID
Rn.95278
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109615.1 NP_001103085.1 6 1108
GenBank mRNA BC131849.1 - 6 1065

View Full Product Details