Gene Transcripts
- Gene Symbol
- Adrb1
- Entrez Gene ID
- 24925
- Gene Name
- adrenoceptor beta 1
- Gene Aliases
- B1AR, RATB1AR
- Chromosome Location
- Chr. 1: 277537585 - 277538985 on Build Rnor_6.0
- UniGene ID
- Rn.87064
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012701.1 |
NP_036833.1
|
1 |
691 |
|
View Full Product Details