Gene Transcripts

Gene Symbol
Ctxn3
Entrez Gene ID
100188934
Gene Name
cortexin 3
Gene Aliases
Chromosome Location
Chr. 18: 52542617 - 52551806 on Build Rnor_6.0
UniGene ID
Rn.115396
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134696.1 NP_001128168.1 3 1189
XM_006254743.2 XP_006254805.1 3 1619
GenBank mRNA BC166550.1 - 958

View Full Product Details