Gene Transcripts
- Gene Symbol
- Mast3
- Entrez Gene ID
- 688540
- Gene Name
- microtubule associated serine/threonine kinase 3
- Gene Aliases
- Chromosome Location
- Chr. 16: 20387338 - 20414767 on Build Rnor_6.0
- UniGene ID
- Rn.155444
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134796.1 |
NP_001128268.1
|
7, 8 |
904 |
|
XM_006252943.2 |
XP_006253005.1
|
8, 9 |
1365 |
|
XM_006252944.2 |
XP_006253006.1
|
8, 9 |
4896 |
|
XM_006252945.2 |
XP_006253007.1
|
9, 10 |
803 |
|
XM_006252946.2 |
XP_006253008.1
|
7, 8 |
4872 |
|
XM_006252947.2 |
XP_006253009.1
|
8, 9 |
782 |
|
XM_008771150.1 |
XP_008769372.1
|
9, 10 |
1386 |
|
XM_008771151.1 |
XP_008769373.1
|
9, 10 |
1383 |
|
XM_008771152.1 |
XP_008769374.1
|
9, 10 |
1371 |
|
XM_008771153.1 |
XP_008769375.1
|
8, 9 |
1362 |
|
XM_008771154.1 |
XP_008769376.1
|
7, 8 |
4872 |
|
View Full Product Details