Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015340.3 | NP_056155.1 | 14 | 1792 | |
XM_005265006.1 | XP_005265063.1 | 13 | 1748 | ||
XM_011533554.2 | XP_011531856.1 | 14 | 1811 | ||
XM_017006042.1 | XP_016861531.1 | 14 | 1811 | ||
GenBank mRNA | AK025465.1 | - | 8 | 1092 | |
AK026113.1 | - | 1 | 15 | ||
AK308146.1 | - | 12 | 1616 | ||
BC025989.2 | - | 14 | 1799 | 4822546 | |
D21851.1 | - | 14 | 1792 |