Gene Transcripts

Gene Symbol
CDK20
Entrez Gene ID
23552
Gene Name
cyclin dependent kinase 20
Gene Aliases
CCRK, CDCH, P42, PNQALRE
Chromosome Location
Chr. 9: 87966441 - 87974780 on Build GRCh38
UniGene ID
Hs.522274
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039803.2 NP_001034892.1 3 580
NM_001170639.1 NP_001164110.1 3 580
NM_001170640.1 NP_001164111.1 3 580
NM_012119.4 NP_036251.2 3 580
NM_178432.3 NP_848519.1 3 619
XM_017014561.1 XP_016870050.1 3 510
XM_017014562.1 XP_016870051.1 3 510
XM_017014563.1 XP_016870052.1 3 510
XM_017014564.1 XP_016870053.1 3 510
GenBank mRNA AF035013.1 - 3 483
AF113130.1 - 3 491
AK075325.1 - 3 372
AK226135.1 - 2 670
AK291713.1 - 3 501
AK294193.1 - 3 333
AK296328.1 - 3 331
AK298993.1 - 3 323
AY904367.1 - 3 246
BC002655.2 - 3 305 3605918
BI757318.1 - 3 431
BX647489.1 - 3 509
CB992644.1 - 3 589
KU178572.1 - 3 246
KU178573.1 - 3 246
KU178574.1 - 3 246
KU178575.1 - 3 246
KU178576.1 - 3 246
KU178577.1 - 3 246

View Full Product Details