Gene Transcripts
- Gene Symbol
- WBP2
- Entrez Gene ID
- 23558
- Gene Name
- WW domain binding protein 2
- Gene Aliases
- GRAMD6, WBP-2
- Chromosome Location
- Chr. 17: 75845699 - 75855420 on Build GRCh38
- UniGene ID
- Hs.514489
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012478.3 |
NP_036610.2
|
1, 2 |
190 |
|
XM_011524580.2 |
XP_011522882.1
|
1, 2 |
111 |
|
GenBank mRNA |
AF087866.1 |
- |
1, 2 |
94 |
|
AK057881.1 |
- |
1, 2 |
95 |
|
AK294082.1 |
- |
1, 2 |
105 |
|
AK294155.1 |
- |
1, 2 |
105 |
|
AK300875.1 |
- |
1, 2 |
105 |
|
BC007452.1 |
- |
1, 2 |
93 |
3140484 |
BC010616.1 |
- |
1, 2 |
103 |
4177319 |
DA278794.1 |
- |
1, 2 |
190 |
|
View Full Product Details