Gene Transcripts

Gene Symbol
PADI4
Entrez Gene ID
23569
Gene Name
peptidyl arginine deiminase 4
Gene Aliases
PAD, PAD4, PADI5, PDI4, PDI5
Chromosome Location
Chr. 1: 17308195 - 17364004 on Build GRCh38
UniGene ID
Hs.522969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB017919.1 - 10 1165
BC025718.1 - 10 1151 5228777

View Full Product Details