Gene Transcripts

Gene Symbol
OPN3
Entrez Gene ID
23596
Gene Name
opsin 3
Gene Aliases
ECPN, PPP1R116
Chromosome Location
Chr. 1: 241593150 - 241640399 on Build GRCh38
UniGene ID
Hs.409081
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014322.2 NP_055137.2 3 1019
GenBank mRNA AF140242.1 - 3 929
AF303588.1 - 3 976
AK310307.1 - 3 502
BC036773.1 - 3 971 5723543
BC051900.1 - 2 573
BC062622.1 - 3 800
BC073872.1 - 2 624

View Full Product Details