Gene Transcripts
- Gene Symbol
- CBY1
- Entrez Gene ID
- 25776
- Gene Name
- chibby family member 1, beta catenin antagonist
- Gene Aliases
- C22orf2, CBY, HS508I15A, PGEA1, PIGEA-14, PIGEA14, arb1
- Chromosome Location
- Chr. 22: 38656653 - 38673850 on Build GRCh38
- UniGene ID
- Hs.334911
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001002880.1 |
NP_001002880.2
|
3, 4 |
356 |
|
NM_015373.3 |
NP_056188.1
|
2, 3 |
216 |
|
GenBank mRNA |
AB111855.1 |
- |
1, 2 |
94 |
|
AF331041.1 |
- |
2, 3 |
152 |
|
AK307265.1 |
- |
2, 3 |
231 |
|
AK311928.1 |
- |
2, 3 |
229 |
|
AL050345.1 |
- |
2, 3 |
195 |
|
AL136686.1 |
- |
2, 3 |
199 |
|
BC016139.1 |
- |
2, 3 |
194 |
3922707 |
CD702742.1 |
- |
2, 3 |
254 |
|
CR456410.1 |
- |
1, 2 |
108 |
|
View Full Product Details