Gene Transcripts

Gene Symbol
POC1A
Entrez Gene ID
25886
Gene Name
POC1 centriolar protein A
Gene Aliases
PIX2, SOFT, WDR51A
Chromosome Location
Chr. 3: 52075233 - 52154690 on Build GRCh38
UniGene ID
Hs.476306
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161580.1 NP_001155052.1 9 1214
NM_001161581.1 NP_001155053.1 9 1285
NM_015426.4 NP_056241.3 9 1214
XM_017006104.1 XP_016861593.1 9 1007
GenBank mRNA AK075289.1 - 9 929
AK315688.1 - 9 936
AL117629.1 - 4 253
BC007417.2 - 8 864
BC110877.1 - 9 1214 5588109
BC119692.2 - 9 924 40116721
BC119693.2 - 9 1000

View Full Product Details