Gene Transcripts
- Gene Symbol
- AHCTF1
- Entrez Gene ID
- 25909
- Gene Name
- AT-hook containing transcription factor 1
- Gene Aliases
- ELYS, MST108, MSTP108, TMBS62
- Chromosome Location
- Chr. 1: 246839098 - 246931978 on Build GRCh38
- UniGene ID
- Hs.300887
- Species
- Homo sapiens
- Gene Symbol
- AHCTF1P1
- Entrez Gene ID
- 285116
- Gene Name
- AT-hook containing transcription factor 1 pseudogene 1
- Gene Aliases
- AHCTF1P, ELYS-like
- Chromosome Location
- Chr. 2: 161499067 - 161507902 on Build GRCh38
- UniGene ID
- Hs.470435
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_077058.1 |
|
1 |
421 |
|
GenBank mRNA |
AK057394.1 |
- |
1 |
255 |
|
View Full Product Details