Gene Transcripts

Gene Symbol
TSKU
Entrez Gene ID
25987
Gene Name
tsukushi, small leucine rich proteoglycan
Gene Aliases
E2IG4, LRRC54, TSK
Chromosome Location
Chr. 11: 76782313 - 76798154 on Build GRCh38
UniGene ID
Hs.8361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258210.1 NP_001245139.1 2 322
NM_001318477.1 NP_001305406.1 2 1219
NM_001318478.1 NP_001305407.1 2 419
NM_001318479.1 NP_001305408.1 2 230
NM_015516.3 NP_056331.2 2 365
GenBank mRNA AF191019.1 - 1 206
AK075476.1 - 2 277
AK091475.1 - 2 278
AK126536.1 - 2 1219
AK313405.1 - 2 278
AY358317.1 - 2 256
BC020975.1 - 2 243 3848274
BT007440.1 - 1 193
DA623614.1 - 2 420
DA879213.1 - 2 230
DB199776.1 - 2 384

View Full Product Details