Gene Transcripts
- Gene Symbol
- SENP3-EIF4A1
- Entrez Gene ID
- 100533955
- Gene Name
- SENP3-EIF4A1 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 17: 7561992 - 7579006 on Build GRCh38
- UniGene ID
- Hs.129673
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037926.1 |
|
9 |
1831 |
|
- Gene Symbol
- SENP3
- Entrez Gene ID
- 26168
- Gene Name
- SUMO1/sentrin/SMT3 specific peptidase 3
- Gene Aliases
- SMT3IP1, SSP3, Ulp1
- Chromosome Location
- Chr. 17: 7561992 - 7571969 on Build GRCh38
- UniGene ID
- Hs.513926
- Species
- Homo sapiens
View Full Product Details