Gene Transcripts

Gene Symbol
OR2K2
Entrez Gene ID
26248
Gene Name
olfactory receptor family 2 subfamily K member 2
Gene Aliases
HSHTPCRH06, HTPCRH06, OR2AN1P, OR2AR1P
Chromosome Location
Chr. 9: 111326884 - 111330224 on Build GRCh38
UniGene ID
Hs.381312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_205859.1 NP_995581.1 1 273
XM_011518520.2 XP_011516822.1 2 441
GenBank mRNA BC111086.1 - 1 273

View Full Product Details