Gene Transcripts

Gene Symbol
OR7A5
Entrez Gene ID
26659
Gene Name
olfactory receptor family 7 subfamily A member 5
Gene Aliases
HTPCR2
Chromosome Location
Chr. 19: 14825743 - 14835197 on Build GRCh38
UniGene ID
Hs.137573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017506.1 NP_059976.1 1 677
XM_011527912.2 XP_011526214.1 2 702
XM_011527913.2 XP_011526215.1 2 628
XM_017026583.1 XP_016882072.1 2 591
GenBank mRNA AK312475.1 - 2 543
BC104809.1 - 1 512 8143812
X64976.1 - 1 80
X89677.1 - 1 80

View Full Product Details