Gene Transcripts

Gene Symbol
CACYBP
Entrez Gene ID
27101
Gene Name
calcyclin binding protein
Gene Aliases
GIG5, PNAS-107, S100A6BP, SIP
Chromosome Location
Chr. 1: 174999435 - 175012027 on Build GRCh38
UniGene ID
Hs.508524
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007214.1 NP_001007215.1 4 625
NM_014412.2 NP_055227.1 4 810
XM_017001046.1 XP_016856535.1 4 757
GenBank mRNA AF057356.1 - 4 499
AF275803.1 - 4 444
AF314752.1 - 4 509
AK093425.1 - 4 597
AK313278.1 - 4 601
AL035305.1 - 4 502
AY423723.1 - 4 386
BC005975.1 - 4 611 4102572
BC022352.1 - 4 508 4281339
BC067823.1 - 4 492 5260416
BC078151.1 - 4 475 6065872
BQ432793.1 - 4 628

View Full Product Details