Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318052.1 | NP_001304981.1 | 2 | 873 | |
NM_001318053.1 | NP_001304982.1 | 3 | 783 | ||
NM_015689.4 | NP_056504.3 | 3 | 975 | ||
NR_134477.1 | 1 | 600 | |||
XM_005249977.3 | XP_005250034.1 | 3 | 1506 | ||
XM_005249979.4 | XP_005250036.1 | 3 | 759 | ||
XM_011516052.2 | XP_011514354.1 | 3 | 853 | ||
XM_011516053.2 | XP_011514355.1 | 2 | 957 | ||
XM_011516054.2 | XP_011514356.1 | 3 | 761 | ||
XM_017011989.1 | XP_016867478.1 | 2 | 959 | ||
GenBank mRNA | AB033103.2 | - | 2 | 873 | |
AK290056.1 | - | 3 | 744 | ||
AK297959.1 | - | 2 | 642 | ||
BC115004.1 | - | 3 | 783 | 7470138 | |
BC132875.1 | - | 1 | 570 | 40146897 | |
DA401015.1 | - | 1 | 315 |