Gene Transcripts
- Gene Symbol
- ERLEC1
- Entrez Gene ID
- 27248
- Gene Name
- endoplasmic reticulum lectin 1
- Gene Aliases
- C2orf30, CIM, CL24936, CL25084, HEL117, XTP3-B, XTP3TPB
- Chromosome Location
- Chr. 2: 53786931 - 53826928 on Build GRCh38
- UniGene ID
- Hs.438336
- Species
- Homo sapiens
- Gene Symbol
- LOC105377134
- Entrez Gene ID
- 105377134
- Gene Name
- uncharacterized LOC105377134
- Gene Aliases
- Chromosome Location
- Chr. 21: 14027399 - 14060948 on Build GRCh38
- UniGene ID
- Hs.558872
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC024173.1 |
- |
3 |
681 |
|
BC048201.1 |
- |
3 |
410 |
|
View Full Product Details