Gene Transcripts
- Gene Symbol
- EML4
- Entrez Gene ID
- 27436
- Gene Name
- echinoderm microtubule associated protein like 4
- Gene Aliases
- C2orf2, ELP120, EMAP-4, EMAPL4, ROPP120
- Chromosome Location
- Chr. 2: 42169338 - 42332548 on Build GRCh38
- UniGene ID
- Hs.713173
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145076.2 |
NP_001138548.1
|
6, 7 |
897 |
|
NM_019063.4 |
NP_061936.2
|
7, 8 |
1071 |
|
XM_005264267.1 |
XP_005264324.1
|
8, 9 |
1084 |
|
XM_005264268.3 |
XP_005264325.1
|
8, 9 |
1084 |
|
XM_006711991.1 |
XP_006712054.1
|
7, 8 |
910 |
|
XM_006711992.2 |
XP_006712055.1
|
9, 10 |
1205 |
|
GenBank mRNA |
AF177377.2 |
- |
7, 8 |
1033 |
|
AK314702.1 |
- |
7, 8 |
1045 |
|
BC140845.1 |
- |
7, 8 |
994 |
9021713 |
BC146799.1 |
- |
6, 7 |
871 |
8860304 |
View Full Product Details